Learn More
Selleck Chemical LLC AT7519 5mg 844442-38-2
SDP
Catalog No. 508820
Encompass_Preferred
AT7519 is a multi-CDK inhibitor for CDK1, 2, 4, 6 and 9 with IC50 of 10-210 nM. It is less potent to CDK3 and little active to CDK7. AT7519 also inhibits GSK3? with IC50 of 89 nM. AT7519 induces apoptosis. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-882-0 Supplier Selleck Chemical LLC Supplier No. S15245MG
AT7519 is a multi-CDK inhibitor for CDK1, 2, 4, 6 and 9 with IC50 of 10-210 nM. It is less potent to CDK3 and little active to CDK7. AT7519 also inhibits GSK3? with IC50 of 89 nM. AT7519 induces apoptosis. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.