Learn More
Selleck Chemical LLC AT7519 HCl 5mg 902135-91-5
SDP
Catalog No. 501365279
Encompass_Preferred
AT7519 HCl is a multi-CDK inhibitor for CDK1, 2, 4, 6 and 9 with IC50 of 10-210 nM in cell-free assays. It is less potent to CDK3 and little active to CDK7. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5279 Supplier Selleck Chemical LLC Supplier No. S78085MG
Add to Cart
Add to Cart
AT7519 HCl is a multi-CDK inhibitor for CDK1, 2, 4, 6 and 9 with IC50 of 10-210 nM in cell-free assays. It is less potent to CDK3 and little active to CDK7. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.