Learn More
Apexbio Technology LLC AT7519 Hydrochloride 902135-91-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000095884
Encompass_Preferred
AT7519 Hydrochloride (CAS 902135-91-5) is a small-molecule inhibitor of cyclin-dependent kinases (CDKs) which are critical regulators of cell proliferation division and apoptosis in eukaryotic cells By targeting multiple CDKs including CDK1 and CDK2 AT7519 induces cell cycle arrest and apoptosis in various human tumor cell lines demonstrating inhibitory activity with IC50 values ranging from 40 to 940 nmol/L In preclinical studies it suppressed tumor growth in xenograft models and promoted tumor cell apoptosis AT7519 is under clinical evaluation for the treatment of refractory solid tumors supporting its utility in cancer research and drug development
Catalog No. 50-000-95884 Supplier Apexbio Technology LLC Supplier No. A31975.1
Add to Cart
Add to Cart
AT7519 Hydrochloride (CAS 902135-91-5) is a small-molecule inhibitor of cyclin-dependent kinases (CDKs) which are critical regulators of cell proliferation division and apoptosis in eukaryotic cells By targeting multiple CDKs including CDK1 and CDK2 AT7519 induces cell cycle arrest and apoptosis in various human tumor cell lines demonstrating inhibitory activity with IC50 values ranging from 40 to 940 nmol/L In preclinical studies it suppressed tumor growth in xenograft models and promoted tumor cell apoptosis AT7519 is under clinical evaluation for the treatment of refractory solid tumors supporting its utility in cancer research and drug development