Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AT7519 Hydrochloride | 902135-91-5 | 99.2% | C16H18Cl3N5O2 | 1 ML
SDP

Catalog No. 5000358289
Encompass_Preferred

AT7519 Hydrochloride is a potent inhibitor of cyclin-dependent kinases (CDKs), with low nanomolar IC50s for CDK1, CDK2, CDK4-CDK6, and CDK9. It induces cytotoxicity and apoptosis in multiple myeloma (MM) cells, linked to GSK-3β activation and inhibition of RNA polymerase II. The compound inhibits tumor growth in MM xenograft mouse models and has been involved in clinical trials for various cancers.

  • Potent inhibitor of CDKs (CDK1, CDK2, CDK4-CDK6, CDK9)
  • Induces cytotoxicity and apoptosis in MM cells
  • Overcomes cytokine and BMSC protective effects
  • Inhibits RNA polymerase II CTD phosphorylation and RNA synthesis
  • Inhibits cell cycle progression and reduces antiapoptotic protein levels
  • Inhibits tumor growth in MM xenograft mouse models

Catalog No. 50-003-58289 Supplier Medchemexpress LLC Supplier No. HY5094310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AT7519 Hydrochloride is a potent inhibitor of cyclin-dependent kinases (CDKs), with low nanomolar IC50s for CDK1, CDK2, CDK4-CDK6, and CDK9. It induces cytotoxicity and apoptosis in multiple myeloma (MM) cells, linked to GSK-3β activation and inhibition of RNA polymerase II. The compound inhibits tumor growth in MM xenograft mouse models and has been involved in clinical trials for various cancers.

  • Potent inhibitor of CDKs (CDK1, CDK2, CDK4-CDK6, CDK9)
  • Induces cytotoxicity and apoptosis in MM cells
  • Overcomes cytokine and BMSC protective effects
  • Inhibits RNA polymerase II CTD phosphorylation and RNA synthesis
  • Inhibits cell cycle progression and reduces antiapoptotic protein levels
  • Inhibits tumor growth in MM xenograft mouse models

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.