Learn More
Medchemexpress LLC HY-50940A 5mg Medchemexpress, AT7519 (trifluoroacetate) CAS:1431697-85-6 Purity:>98%
SDP
Catalog No. 502082261
Encompass_Preferred
Medchemexpress, HY-50940A 5mg AT7519 (trifluoroacetate) CAS:1431697-85-6 AT7519 trifluoroacetate as a potent inhibitor of CDKs, with IC50s of 210, 47, 100, 13, 170, and <10 nM for CDK1, CDK2, CDK4 to CDK6, and CDK9, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2261 Supplier Medchemexpress LLC Supplier No. HY50940A5MG
Medchemexpress, HY-50940A 5mg AT7519 (trifluoroacetate) CAS:1431697-85-6 AT7519 trifluoroacetate as a potent inhibitor of CDKs, with IC50s of 210, 47, 100, 13, 170, and <10 nM for CDK1, CDK2, CDK4 to CDK6, and CDK9, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.