Learn More
Cayman Chemical AT7519hydrochlorIde 10mg
SDP
Catalog No. 502972986
Encompass_Preferred
An ATP-competitive inhibitor of Cdk1, 2, 4, 5, 6, and 9 with IC50 values of 210, 47, 100, 13, 170, and 50s = 40-940 nM in vitro) and human tumor xenograft models, inhibiting cell cycle progression and inducing apoptosis
Catalog No. 50-297-2986 Supplier Cayman Chemical Supplier No. 1623110 MG
An ATP-competitive inhibitor of Cdk1, 2, 4, 5, 6, and 9 with IC50 values of 210, 47, 100, 13, 170, and 50s = 40-940 nM in vitro) and human tumor xenograft models, inhibiting cell cycle progression and inducing apoptosis