Learn More
Medchemexpress LLC HY-12059 5mg Medchemexpress, AT7867 CAS:857531-00-1 Purity:>98%
SDP
Catalog No. 502029887
Encompass_Preferred
Medchemexpress, HY-12059 5mg AT7867 CAS:857531-00-1 AT7867 is a potent ATP-competitive inhibitor of Akt1/Akt2/Akt3 and p70S6K/PKA with IC50s of 32 nM/17 nM/47 nM and 85 nM/20 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9887 Supplier Medchemexpress LLC Supplier No. HY120595MG
Medchemexpress, HY-12059 5mg AT7867 CAS:857531-00-1 AT7867 is a potent ATP-competitive inhibitor of Akt1/Akt2/Akt3 and p70S6K/PKA with IC50s of 32 nM/17 nM/47 nM and 85 nM/20 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.