Learn More
Medchemexpress LLC HY-12059A 5mg Medchemexpress, AT7867 (dihydrochloride) CAS:1431697-86-7 Purity:>98%
SDP
Catalog No. 502080944
Encompass_Preferred
Medchemexpress, HY-12059A 5mg AT7867 (dihydrochloride) CAS:1431697-86-7 AT7867 dihydrochloride is a potent ATP-competitive inhibitor of Akt1/Akt2/Akt3 and p70S6K/PKA with IC50s of 32 nM/17 nM/47 nM and 85 nM/20 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0944 Supplier Medchemexpress LLC Supplier No. HY12059A5MG
Add to Cart
Add to Cart
Medchemexpress, HY-12059A 5mg AT7867 (dihydrochloride) CAS:1431697-86-7 AT7867 dihydrochloride is a potent ATP-competitive inhibitor of Akt1/Akt2/Akt3 and p70S6K/PKA with IC50s of 32 nM/17 nM/47 nM and 85 nM/20 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.