Learn More
Selleck Chemical LLC AT9283 S1134-2mg
SDP
Catalog No. 503108365
Encompass_Preferred
AT9283 is a potent JAK2/3 inhibitor with IC50 of 1 2 nM/1 1 nM in cell-free assays also potent to Aurora A/B Abl1(T315I)
Catalog No. 50-310-8365 Supplier Selleck Chemical LLC Supplier No. S11342MG
AT9283 is a potent JAK2/3 inhibitor with IC50 of 1 2 nM/1 1 nM in cell-free assays also potent to Aurora A/B Abl1(T315I)