Learn More
Apexbio Technology LLC AT9283 896466-04-9 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A41175.1
Encompass_Preferred
AT9283 (CAS 896466-04-9) is a synthetic small-molecule inhibitor developed through fragment-based drug design It inhibits Aurora kinases A and B key components of the serine/threonine kinase family involved in regulating cell division processes as well as targets including Janus kinases (JAKs) BCR-ABL kinase (including the T315I mutant) and Flt-3 With a potent IC50 around 3 nM against Aurora kinases AT9283 exhibits antiproliferative and pro-apoptotic activity in leukemia myeloproliferative diseases solid tumors and B-cell lymphoma models It is utilized primarily in cancer biology research to investigate its therapeutic potential targeting aberrant kinase activity
Catalog No. 50-000-62637