Learn More
Medchemexpress LLC AT9283 | 896466-04-9 | 99.7% | C19H23N7O2 | 200 MG
SDP
Catalog No. 5000358811
Encompass_Preferred
AT9283 is a multi-targeted kinase inhibitor with potent activity against Aurora A/B, JAK2/3, Abl (T315I), and Flt3 (IC50s 1-30 nM). It has been shown to inhibit the growth and survival of various solid tumors both in vitro and in vivo. This product is intended for research use only.
  • Potent multi-targeted kinase inhibitor
  • Active against Aurora A/B, JAK2/3, Abl (T315I), and Flt3
  • Inhibits growth and survival of various solid tumors
  • Induces polyploid phenotype and apoptosis in certain cancer cell lines
  • Shows significant tumor growth inhibition in xenograft models
  • Enhances survival in mouse xenograft models of mantle cell lymphoma
Catalog No. 50-003-58811 Supplier Medchemexpress LLC Supplier No. HY50514200MG
AT9283 is a multi-targeted kinase inhibitor with potent activity against Aurora A/B, JAK2/3, Abl (T315I), and Flt3 (IC50s 1-30 nM). It has been shown to inhibit the growth and survival of various solid tumors both in vitro and in vivo. This product is intended for research use only.
  • Potent multi-targeted kinase inhibitor
  • Active against Aurora A/B, JAK2/3, Abl (T315I), and Flt3
  • Inhibits growth and survival of various solid tumors
  • Induces polyploid phenotype and apoptosis in certain cancer cell lines
  • Shows significant tumor growth inhibition in xenograft models
  • Enhances survival in mouse xenograft models of mantle cell lymphoma