Learn More
Selleck Chemical LLC AT9283 5mg 896466-04-9
SDP
Catalog No. 506232
Encompass_Preferred
AT9283 is a potent JAK2/3 inhibitor with IC50 of 1.2 nM/1.1 nM in cell-free assays; also potent to Aurora A/B, Abl1(T315I). Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-623-2 Supplier Selleck Chemical LLC Supplier No. S11345MG
AT9283 is a potent JAK2/3 inhibitor with IC50 of 1.2 nM/1.1 nM in cell-free assays; also potent to Aurora A/B, Abl1(T315I). Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.