Learn More
Medchemexpress LLC HY-109052 5mg Medchemexpress, Atabecestat CAS:1200493-78-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1090525MG
Encompass_Preferred
Medchemexpress, HY-109052 5mg Atabecestat CAS:1200493-78-2 Atabecestat (JNJ-54861911) is a potent brain-penetrant and orally active β-site amyloid precursor protein cleaving enzyme 1 (BACE1) inhibitor, achieves robust and high CSF Aβ reduction. Atabecestat (JNJ-54861911) is tolerated and displays a sustained pharmacokinetic (PK) and pharmacodynamic (PD) characteristics. Atabecestat (JNJ-54861911) has the potential for Alzheimer's Disease treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9661