Learn More
Active Motif Active Motif ATAC-Seq Spike-In Control, Reduces Technical Variation, Normalizes ATAC-Seq Data, 16 Reactions
SDP
Catalog No. NC2881031
Encompass
The Active Motif ATAC-Seq Spike-In Control enhances experimental accuracy by utilizing cryopreserved *Drosophila* cell nuclei for normalization in ATAC-Seq workflows. This innovative control minimizes technical variations and reduces sample processing bias, leading to more reliable sequencing results. With a reaction quantity of 16 reactions, this product stands out by ensuring consistent tagging across open chromatin regions, making it essential for precise data analysis.
Catalog No. NC2881031 Supplier Active Motif Supplier No. 53154
The Active Motif ATAC-Seq Spike-In Control enhances experimental accuracy by utilizing cryopreserved *Drosophila* cell nuclei for normalization in ATAC-Seq workflows. This innovative control minimizes technical variations and reduces sample processing bias, leading to more reliable sequencing results. With a reaction quantity of 16 reactions, this product stands out by ensuring consistent tagging across open chromatin regions, making it essential for precise data analysis.