Learn More
Medchemexpress LLC Ataciguat | 254877-67-3 | MFCD09837865 | 99.7% | 576.49 g·mol⁻1 | C21H19Cl2N3O6S3 | 10 MG
SDP
Catalog No. 5000298353
Encompass_Preferred
Ataciguat (HMR-1766) is a nitric oxide-independent soluble guanylate cyclase (sGC) activator used in preclinical research to stimulate cyclic GMP (cGMP) production and induce vasodilation. It is supplied in solid forms and as a DMSO solution for use in in vitro and in vivo studies.
  • Activates the ferric heme-iron redox form of sGC to increase cGMP levels.
  • Exhibits vasodilator effects relevant to cardiovascular research.
  • Available as multiple solid sizes and a 10 mM DMSO solution for flexible dosing.
  • High reported purity (99.7%) for consistent experimental results.
  • Molecular weight 576.49 g·mol⁻1 and formula C21H19Cl2N3O6S3 for accurate identification.
Catalog No. 50-002-98353 Supplier Medchemexpress LLC Supplier No. HY1750010MG
Ataciguat (HMR-1766) is a nitric oxide-independent soluble guanylate cyclase (sGC) activator used in preclinical research to stimulate cyclic GMP (cGMP) production and induce vasodilation. It is supplied in solid forms and as a DMSO solution for use in in vitro and in vivo studies.
  • Activates the ferric heme-iron redox form of sGC to increase cGMP levels.
  • Exhibits vasodilator effects relevant to cardiovascular research.
  • Available as multiple solid sizes and a 10 mM DMSO solution for flexible dosing.
  • High reported purity (99.7%) for consistent experimental results.
  • Molecular weight 576.49 g·mol⁻1 and formula C21H19Cl2N3O6S3 for accurate identification.