Learn More
Cole-Parmer Atago 3810 (PAL-1) Digital Pocket Refractometer, 0-53% Brix
Supplier:  Cole-Parmer 0294151
Encompass_Preferred
Compact, durable unit provides simple and accurate Brix readings
Lightweight and compact pocket refractometers are ideal for determining Brix concentrations in fruits and beverages, cutting oils, and organic samples. Use in the field or indoors, measurements are quick and accurate.
Catalog No. NC1790740