Learn More
Selleck Chemical LLC Ataluren (PTC124) 10mM/1mL 775304-57-9
SDP
Catalog No. 501364308
Encompass_Preferred
Ataluren (PTC124) selectively induces ribosomal read-through of premature but not normal termination codons, with EC50 of 0.1 ?M in HEK293 cells, may provide treatment for genetic disorders caused by nonsense mutations (e.g. CF caused by CFTR nonsense mutation). Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4308 Supplier Selleck Chemical LLC Supplier No. S600310MM/1ML
Ataluren (PTC124) selectively induces ribosomal read-through of premature but not normal termination codons, with EC50 of 0.1 ?M in HEK293 cells, may provide treatment for genetic disorders caused by nonsense mutations (e.g. CF caused by CFTR nonsense mutation). Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.