Learn More
Selleck Chemical LLC Atamparib S8993-25MG
SDP
Catalog No. 5000023685
Encompass_Preferred
Atamparib (RBN-2397) is a potent selective and orally active NAD competitive inhibitor PARP7 with IC50 of 3 nM and Kd of 0 001 M RBN-2397 has the potential for the research of tumor treatment
Catalog No. 50-000-23685 Supplier Selleck Chemical LLC Supplier No. S899325MG
Atamparib (RBN-2397) is a potent selective and orally active NAD competitive inhibitor PARP7 with IC50 of 3 nM and Kd of 0 001 M RBN-2397 has the potential for the research of tumor treatment