Learn More
Aviva Systems Biology Corp Atat1 Antibody - middle region (ARP42644_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42644_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Atat1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Rat Atat1. Alternate names: 3110080J08Rik, FLJ13158|Mec17|TATApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-014