Learn More
Apexbio Technology LLC Atazanavir 198904-31-3 5mg
SDP
Catalog No. 5000092096
Encompass_Preferred
Atazanavir (CAS 198904-31-3) is a small-molecule inhibitor targeting HIV-1 protease It is designed to block protease-mediated cleavage of viral gag and gag-pol polyproteins thereby preventing the maturation of infectious viral particles Atazanavir exerts its biological activity primarily through direct binding to the active site of HIV-1 protease inhibiting polyprotein cleavage In in vitro studies atazanavir demonstrates inhibition of HIV-1 replication with reported EC50 values ranging from 2 6 to 5 3 nmol/L and a protease-inhibition constant (Ki) of approximately 66 nmol/L Based on these pharmacological properties atazanavir holds research potential in anti-retroviral assays and studies involving HIV drug resistance and protease inhibition profiling
Catalog No. 50-000-92096 Supplier Apexbio Technology LLC Supplier No. A82055
Atazanavir (CAS 198904-31-3) is a small-molecule inhibitor targeting HIV-1 protease It is designed to block protease-mediated cleavage of viral gag and gag-pol polyproteins thereby preventing the maturation of infectious viral particles Atazanavir exerts its biological activity primarily through direct binding to the active site of HIV-1 protease inhibiting polyprotein cleavage In in vitro studies atazanavir demonstrates inhibition of HIV-1 replication with reported EC50 values ranging from 2 6 to 5 3 nmol/L and a protease-inhibition constant (Ki) of approximately 66 nmol/L Based on these pharmacological properties atazanavir holds research potential in anti-retroviral assays and studies involving HIV drug resistance and protease inhibition profiling