Learn More
Medchemexpress LLC Atazanavir | 198904-31-3 | MFCD09839958 | >98% | 704.86 | 50 MG
SDP
Catalog No. 5000385202
Encompass_Preferred
Atazanavir (BMS-232632) is a highly selective and orally active HIV-1 protease inhibitor. Atazanavir is a substrate and inhibitor of CYP3A4, and an inhibitor of P-glycoprotein (P-gp). Atazanavir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 3.49 μM. Atazanavir inhibits cardiac fibrosis, hyperlipidemia and induces malignant glioma death.
  • HIV-1 protease inhibitor.
  • Substrate and inhibitor of CYP3A4.
  • Inhibitor of P-glycoprotein (P-gp).
  • SARS-CoV 3CLpro inhibitor (IC50 = 3.49 μM).
  • Inhibits cardiac fibrosis and hyperlipidemia.
  • Induces malignant glioma death.
Catalog No. 50-003-85202 Supplier Medchemexpress LLC Supplier No. HY1736750MG
Atazanavir (BMS-232632) is a highly selective and orally active HIV-1 protease inhibitor. Atazanavir is a substrate and inhibitor of CYP3A4, and an inhibitor of P-glycoprotein (P-gp). Atazanavir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 3.49 μM. Atazanavir inhibits cardiac fibrosis, hyperlipidemia and induces malignant glioma death.
  • HIV-1 protease inhibitor.
  • Substrate and inhibitor of CYP3A4.
  • Inhibitor of P-glycoprotein (P-gp).
  • SARS-CoV 3CLpro inhibitor (IC50 = 3.49 μM).
  • Inhibits cardiac fibrosis and hyperlipidemia.
  • Induces malignant glioma death.