Learn More
Medchemexpress LLC Atazanavir-d6 | 1092540-50-5 | 99.3% | 710.89 | 500 UG
SDP
Catalog No. 5000384476
Encompass_Preferred
Atazanavir-d6 is deuterium labeled Atazanavir. Atazanavir (BMS-232632) is a highly selective HIV-1 protease inhibitor and the first protease inhibitor approved for once-daily administration. It is a substrate and inhibitor of CYP3A4, and an inhibitor and inducer of P-glycoprotein (P-gp). Atazanavir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 3.49 μM.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-84476 Supplier Medchemexpress LLC Supplier No. HY17367S4500UG
Atazanavir-d6 is deuterium labeled Atazanavir. Atazanavir (BMS-232632) is a highly selective HIV-1 protease inhibitor and the first protease inhibitor approved for once-daily administration. It is a substrate and inhibitor of CYP3A4, and an inhibitor and inducer of P-glycoprotein (P-gp). Atazanavir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 3.49 μM.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.