Learn More
Medchemexpress LLC Atazanavir sulfate | 229975-97-7 | 802.93 | 1 ML
SDP
Catalog No. 5000385530
Encompass_Preferred
Atazanavir (BMS-232632) is a highly selective and orally active HIV-1 protease inhibitor. It acts as a substrate and inhibitor of CYP3A4, and an inhibitor of P-glycoprotein (P-gp). Atazanavir also inhibits SARS-CoV 3CL pro with an IC50 of 3.49 μM. Additionally, Atazanavir has been shown to inhibit cardiac fibrosis, hyperlipidemia, and induce malignant glioma cell death.
  • Highly selective HIV-1 protease inhibitor
  • Orally active
  • Inhibits CYP3A4
  • Inhibits P-glycoprotein (P-gp)
  • Inhibits SARS-CoV 3CL pro
  • Inhibits cardiac fibrosis
  • Inhibits hyperlipidemia
  • Induces malignant glioma death
Catalog No. 50-003-85530 Supplier Medchemexpress LLC Supplier No. HY1736710MM/1ML
Atazanavir (BMS-232632) is a highly selective and orally active HIV-1 protease inhibitor. It acts as a substrate and inhibitor of CYP3A4, and an inhibitor of P-glycoprotein (P-gp). Atazanavir also inhibits SARS-CoV 3CL pro with an IC50 of 3.49 μM. Additionally, Atazanavir has been shown to inhibit cardiac fibrosis, hyperlipidemia, and induce malignant glioma cell death.
  • Highly selective HIV-1 protease inhibitor
  • Orally active
  • Inhibits CYP3A4
  • Inhibits P-glycoprotein (P-gp)
  • Inhibits SARS-CoV 3CL pro
  • Inhibits cardiac fibrosis
  • Inhibits hyperlipidemia
  • Induces malignant glioma death