Learn More
Medchemexpress LLC Atazanavir (sulfate) | 229975-97-7 | 99.7% | 802.93 | 50 MG
SDP
Catalog No. 5000384565
Encompass_Preferred
Atazanavir sulfate is a highly selective and orally active HIV-1 protease inhibitor with blood-brain barrier permeability. It functions as both a substrate and inhibitor of CYP3A4, and also inhibits P-glycoprotein (P-gp). Additionally, it is a SARS-CoV 3CLpro inhibitor with an IC50 of 3.49 μM. This compound has demonstrated the ability to inhibit cardiac fibrosis and hyperlipidemia, and induce malignant glioma cell death.
  • Highly selective and orally active
  • HIV-1 protease inhibitor
  • Crosses the blood-brain barrier
  • Inhibitor and substrate of CYP3A4
  • Inhibits P-glycoprotein (P-gp)
  • SARS-CoV 3CLpro inhibitor (IC50 of 3.49 μM)
  • Inhibits cardiac fibrosis
  • Inhibits hyperlipidemia
  • Induces malignant glioma cell death
Catalog No. 50-003-84565 Supplier Medchemexpress LLC Supplier No. HY17367A50MG
Atazanavir sulfate is a highly selective and orally active HIV-1 protease inhibitor with blood-brain barrier permeability. It functions as both a substrate and inhibitor of CYP3A4, and also inhibits P-glycoprotein (P-gp). Additionally, it is a SARS-CoV 3CLpro inhibitor with an IC50 of 3.49 μM. This compound has demonstrated the ability to inhibit cardiac fibrosis and hyperlipidemia, and induce malignant glioma cell death.
  • Highly selective and orally active
  • HIV-1 protease inhibitor
  • Crosses the blood-brain barrier
  • Inhibitor and substrate of CYP3A4
  • Inhibits P-glycoprotein (P-gp)
  • SARS-CoV 3CLpro inhibitor (IC50 of 3.49 μM)
  • Inhibits cardiac fibrosis
  • Inhibits hyperlipidemia
  • Induces malignant glioma cell death