Learn More
Apexbio Technology LLC Atazanavir sulfate (BMS-232632-05) 229975-97-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000096728
Encompass_Preferred
Atazanavir sulfate (BMS-232632-05) (CAS 229975-97-7) is a small-molecule inhibitor targeting HIV-1 protease It is designed to inhibit protease-mediated processing of HIV gag and gag-pol polyproteins thereby blocking viral particle maturation Atazanavir sulfate exerts its biological activity primarily through selective inhibition of HIV-1 protease In in vitro studies atazanavir sulfate demonstrates inhibitory activity against HIV-1 strains with reported IC50 values ranging from 2 to 5 nM Based on these pharmacological properties atazanavir sulfate holds research potential in antiviral studies focused on HIV replication mechanisms and investigations of drug metabolism mediated by hepatic CYP systems
Catalog No. 50-000-96728 Supplier Apexbio Technology LLC Supplier No. A40405.1
Atazanavir sulfate (BMS-232632-05) (CAS 229975-97-7) is a small-molecule inhibitor targeting HIV-1 protease It is designed to inhibit protease-mediated processing of HIV gag and gag-pol polyproteins thereby blocking viral particle maturation Atazanavir sulfate exerts its biological activity primarily through selective inhibition of HIV-1 protease In in vitro studies atazanavir sulfate demonstrates inhibitory activity against HIV-1 strains with reported IC50 values ranging from 2 to 5 nM Based on these pharmacological properties atazanavir sulfate holds research potential in antiviral studies focused on HIV replication mechanisms and investigations of drug metabolism mediated by hepatic CYP systems