Learn More
Selleck Chemical LLC Atazanavir Sulfate 10mg 229975-97-7 BMS-232632
SDP
Catalog No. 507526
Encompass_Preferred
Atazanavir Sulfate (BMS-232632) is a HIV protease inhibitor with Ki of 2.66 nM in a cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-752-6 Supplier Selleck Chemical LLC Supplier No. S145710MG
Atazanavir Sulfate (BMS-232632) is a HIV protease inhibitor with Ki of 2.66 nM in a cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.