Learn More
Medchemexpress LLC HY-15028 10mg Medchemexpress, ATB-346 CAS:1226895-20-0 Purity:>98%
SDP
Catalog No. 502081673
Encompass_Preferred
Medchemexpress, HY-15028 10mg ATB-346 CAS:1226895-20-0 ATB-346 an orally active non-steroidal anti-inflammatory drug (NSAID), inhibits cyclooxygenase-1 and 2 (COX-1 and 2) ATB-346 possesses antiinflammatory and antinociceptive activities[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1673 Supplier Medchemexpress LLC Supplier No. HY1502810MG
Medchemexpress, HY-15028 10mg ATB-346 CAS:1226895-20-0 ATB-346 an orally active non-steroidal anti-inflammatory drug (NSAID), inhibits cyclooxygenase-1 and 2 (COX-1 and 2) ATB-346 possesses antiinflammatory and antinociceptive activities[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.