Learn More
TARGETMOL CHEMICALS INC ATC 0175 HYDROCHLORIDE 5MG
SDP
Catalog No. 502580943
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. ATC 0175 hydrochloride is an orally active melanocyte concentrating hormone 1 receptor antagonist that is potent and selective. ATC 0175 hydrochloride has affinity for MCH1R MCH2R MCH1 5-HT2B receptors and 5-HT1A receptors with IC50s of 13.5 nM >10000 nM 13 nM 9.66 nM and 16.9 nM respectively. ATC 0175 hydrochloride exhibits antidepressant and anxiolytic effects in animal models. aTC 0175 can be used to study depression and/or anxiety disorders. purity: 99%
Catalog No. 50-258-0943 Supplier TARGETMOL CHEMICALS INC Supplier No. T225915MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. ATC 0175 hydrochloride is an orally active melanocyte concentrating hormone 1 receptor antagonist that is potent and selective. ATC 0175 hydrochloride has affinity for MCH1R MCH2R MCH1 5-HT2B receptors and 5-HT1A receptors with IC50s of 13.5 nM >10000 nM 13 nM 9.66 nM and 16.9 nM respectively. ATC 0175 hydrochloride exhibits antidepressant and anxiolytic effects in animal models. aTC 0175 can be used to study depression and/or anxiety disorders. purity: 99%