Learn More
Aviva Systems Biology Corp ATCAY Antibody - C-terminal region (ARP60863_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60863_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATCAY. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ATCAY. Alternate names: BNIP-H, CLAC, KIAA1872ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rat, Yeast
Catalog No. 50-287-191