Learn More
ATCC ATCC B Lymphocyte Cell Line, Homo Sapiens, Burkitt's Lymphoma, BSL 2
Supplier: ATCC CRL2392
This B lymphocyte cell line, isolated from a 41-year-old White male with Burkitt's lymphoma, is intended for immunology research. It expresses high levels of CD77 (approx. 90%) and exhibits sensitivity to Shiga Toxin (Stx). Epstein-Barr virus (EBV) has been detected in this cell line.
- Biosafety level: BSL 2
- Species: Homo sapiens
- Cell type: B lymphocyte, lymphoblast; morphology lymphoblast
- Disease: Burkitt's lymphoma
- Growth properties: Suspension
- Derived from lymphoma cells of a 41-year-old White male
- Antigen expression: CD10+, CD19+, CD20+, CD40+ (10%), CD45+, CD77+ (90%), CD34–
- Sensitive to Shiga Toxin (Stx)
- Epstein-Barr virus (EBV) detected
- Frozen product format for storage in vapor phase of liquid nitrogen
- Population doubling time: Approx. 24 hr.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.