Learn More
ATCC ThawReady™ THP-1; 10 pack
Catalog No. 5000550107
Encompass
ATCC ThawReady™THP-1 monocytes can be used at thaw, eliminating lengthy cell expansion. These assay ready cells allow you to acquire your data in hours rather than weeks, eliminating the need for cell expansion or banking. 10 vial pack.
Catalog No. 50-005-50107 Supplier ATCC Supplier No. TIB202AR10
ATCC ThawReady™THP-1 monocytes can be used at thaw, eliminating lengthy cell expansion. These assay ready cells allow you to acquire your data in hours rather than weeks, eliminating the need for cell expansion or banking. 10 vial pack.