Learn More
ATCC ThawReady™ THP-1 NF-kB-Luc2; 3-Pack
Catalog No. 5000550366
Encompass
ATCC ThawReady™ THP-1 NF-kB-Luc2 monocytes can be used at thaw, eliminating lengthy cell expansion. These assay ready cells allow you to acquire your data in hours rather than weeks, eliminating the need for cell expansion or banking. Additionally, these cells express luciferase under the control of the NF-kB reporter, allowing sensitive and quantitative quantification of NF-kB activation. 3 vial pack.
Catalog No. 50-005-50366 Supplier ATCC Supplier No. TIB202NFLUC2AR3
ATCC ThawReady™ THP-1 NF-kB-Luc2 monocytes can be used at thaw, eliminating lengthy cell expansion. These assay ready cells allow you to acquire your data in hours rather than weeks, eliminating the need for cell expansion or banking. Additionally, these cells express luciferase under the control of the NF-kB reporter, allowing sensitive and quantitative quantification of NF-kB activation. 3 vial pack.