Learn More
Electron Microscopy Sciences Autoclavable Biohazard Bag Red, 8 x 12"

Supplier: Electron Microscopy Sciences 6442102
These autoclavable polyprolyene biohazard bags provide excellent heat and are tear and puncture resistance. They can withstand autoclave temperatures up to 147 deg C (297 deg F). All bags are marked with the standard OSHA symbol and warnings. Indicator strip changes color after an effective autoclave cycle is completed (127 deg C; 260 deg F for 25 minutes).
Color: Red
Size: 8 x 12"
Thickness: 0.0018"
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.