Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens™ Atellica™ CH 930 Analyzer

Catalog No. 23313251 Shop All Siemens Healthineers Products
Change view
Click to view available options
Sample Type:
Whole Blood
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
23-313-251 Whole Blood
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-313-251 Supplier Siemens™ Supplier No. 11067000

This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Atellica™ CH 930 Analyzer uses proven micro-volume technology for photometric testing and highly reliable integrated multisensor technology (IMT) for electrolyte testing.

  • Uses the same reagents and consumables in every configuration for streamlined inventory management and consistent patient results, no matter where samples are tested
  • Up to six Atellica™ CH 930 Analyzers can be connected in the Atellica™ Solution to accommodate chemistry testing volume
  • Up to 1800 Tests/hr. (1200 Tests/hr. Photometric, 600 Tests/hr. IMT)
TRUSTED_SUSTAINABILITY

Specifications

Type Clinical Chemistry Analyzer
Sample Type Whole Blood
Throughput ≤1800 Tests/hr.
Analytical Method Photometric, Turbometric, Ion-selective Electrode
Capacity (Metric) 4 to 50 μL
Reagent Capacity 140 Positions
Cuvettes Yes
Depth (Metric) 115.6 cm
Height (Metric) 136.4 cm
Width (Metric) 149.1 cm
Voltage 200/240 V
Measurement Method Electrolyte (IMT), Photometric
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.