Learn More
Siemens Healthineers™ Atellica™ Decapper
Catalog No. 23313024 Shop All Siemens Healthineers Products
Change view
Click to view available options
Sample Type:
Blood
Catalog No. Sample Type
23-313-024 Blood
1 options
This item is not returnable. View return policy
Catalog No. 23-313-024 Supplier Siemens Healthineers™ Supplier No. 11069400
This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.
This item is not returnable. View return policy
Automation that can be 'added' to Atellica™ CH and IM Analyzers.
  • Automated scheduling and delivery of controls and calibrators to all connected analyzers from an onboard refrigerated compartment
  • Decapping integrated into the analytical workflow of Atellica™ Solution1 with little to no additional footprint
  • Analyzers can be combined into over 300 customizable configurations including linear, L, and U shapes, providing the flexibility and scalability to meet changing testing demands
  • Can be configured as a stand-alone system and is automation-ready to connect to Aptio™ Automation, providing a multidisciplinary total lab solution
  • Same reagents and consumables used across configurations and locations for streamlined inventory control and consistent patient results, no matter where samples are tested
  • More than 30 different sample containers can be used, including pediatric, tube-top cups, and special containers, reducing the number of pour-offs, special handling, and material costs
  • Broad and expanding menu across disease states with proven detection technologies to inform clinical decisions
  • High productivity per square meter with an immunoassay analyzer that runs up to 440 tests/hr.
  • Service innovations, including the Guardian ProgramSM and the Intelligent Service Assistant, to maintain system operations at peak performance
  • Operator tablet provides system monitoring and control and access to live chat, voice, and video remote support via Remote Assistance and built-in system wireless capability
  • Atellica Asa™ Application—your trusted assistant—organizes and prioritizes operator tasks for Atellica™ Solution analyzers by streamlining laboratory workflow, allowing you to confidently walk away from your analyzer and return when intervention is needed
TRUSTED_SUSTAINABILITY
Specifications
Type Analyzer Accessory
Sample Type Blood
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov