Learn More
Siemens Healthineers™ Atellica™ Direct Load System
Catalog No. 23313021 Shop All Siemens Healthineers Products
Change view
Click to view available options
Sample Type:
Blood
Catalog No. Sample Type
23-313-021 Blood
1 options
This item is not returnable. View return policy
Catalog No. 23-313-021 Supplier Siemens Healthineers™ Supplier No. 11068008
This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.
This item is not returnable. View return policy
Multi-camera vision system with 360° view designed to reduce bar-code reading and other errors.
  • 360° view also reduces exception handling and operator intervention
TRUSTED_SUSTAINABILITY
Specifications
Type Analyzer Camera
Sample Type Blood
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov