Learn More
Siemens Healthineers™ Atellica™ IM 1600 Analyzer
Catalog No. 23313018 Shop All Siemens Healthineers Products
Change view
Click to view available options
Sample Type:
Blood
Catalog No. Sample Type
23-313-018 Blood
1 options
This item is not returnable. View return policy
Catalog No. 23-313-018 Supplier Siemens Healthineers™ Supplier No. 11066000
This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.
This item is not returnable. View return policy
The Atellica™ line provides flexible, scalable, automation-ready immunoassay and clinical chemistry analyzers featuring patented, bidirectional magnetic sample transport technology and the flexibility to create over 300 customizable configurations.
  • Includes broad assay menu with proven detection technologies
  • Independent control over every sample impacts operator intervention and turnaround time
  • Patented Atellica™ Magline™ Transport provides rapid, bidirectional, variable-speed transport of every sample, including STATs, pediatric, and special sample containers
TRUSTED_SUSTAINABILITY
Specifications
Type Analyzer
Sample Type Blood
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov