Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atenolol-d7, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012744 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2744 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2744 Supplier MilliporeSigma Supelco Supplier No. 066132MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706030, T706025

Featured Industries: Environmental Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 1202864-50-3
Linear Formula C14D7H15N2O3
MDL Number MFCD06658802
Synonym 4-[2-Hydroxy-3-[(1-methylethyl-d7)amino]propoxy]benzeneacetamide
Type Analytical Standard
Quantity 2 mg
Formula Weight 273.38
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.