Learn More
Cayman Chemical aTerpnene 50mg
SDP
Catalog No. 502964094
Encompass_Preferred
A dopamine D2 receptor antagonist (Ki 0 3 nM in CHO cells expressing the rat receptor) selective for dopamine D2 over D3 receptors (Ki 9 5 nM) inhibits dipropyl dopamine-induced femoral vasodilation in dogs at 0 5-5 g/kg prevents dopamine-induced decreases in pentagastrin-induced gastric antral motility in dogs at 0 5 mg/kg inhibits apomorphine-induced emesis in dogs (ED50 0 031 mg/kg p o ) increases serum levels of prolactin in male rats
Catalog No. 50-296-4094 Supplier Cayman Chemical Supplier No. 23178 50MG
A dopamine D2 receptor antagonist (Ki 0 3 nM in CHO cells expressing the rat receptor) selective for dopamine D2 over D3 receptors (Ki 9 5 nM) inhibits dipropyl dopamine-induced femoral vasodilation in dogs at 0 5-5 g/kg prevents dopamine-induced decreases in pentagastrin-induced gastric antral motility in dogs at 0 5 mg/kg inhibits apomorphine-induced emesis in dogs (ED50 0 031 mg/kg p o ) increases serum levels of prolactin in male rats