Learn More
Medchemexpress LLC HY-P9904 5mg Medchemexpress, Atezolizumab CAS:1380723-44-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99045MG
Encompass_Preferred
Medchemexpress, HY-P9904 5mg Atezolizumab CAS:1380723-44-3 Atezolizumab is a selective humanized monoclonal IgG1 antibody against programmed death ligand 1 ( PD-L1 ), used for cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4393