Learn More
Aviva Systems Biology Corp ATF1 Antibody - middle region (ARP31366_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31366_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ATF1. Alternate names: EWS-ATF1, FUS/ATF-1, TREB36ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54197