Learn More
Aviva Systems Biology Corp ATF2 Antibody - C-terminal region (ARP34151_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34151_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ATF2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human ATF2. Alternate names: HB16, CREB2, TREB7, CRE-BP1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55272