Learn More
Aviva Systems Biology Corp Atf4 Antibody - N-terminal region (ARP38066_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38066_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Atf4. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Atf-4, C/ATF, CREB2, MGC96460, TAXREB67ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Sheep
Catalog No. 50-284-710