Learn More
Aviva Systems Biology Corp ATF4 Antibody - middle region (P100913_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100913_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATF4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ATF4. Alternate names: CREB-2, CREB2, TAXREB67, TXREBApplicationWB. Species reactivity: Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-62781