Learn More
Aviva Systems Biology Corp ATF4P3 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP51150_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC643159. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human LOC643159. Alternate names: ATF4CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Rat, Sheep
Catalog No. 50-285-559