Learn More
Aviva Systems Biology Corp Atf5 Antibody - C-terminal region (ARP57978_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57978_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Atf5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Rat Atf5. Alternate names: -ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-286-423