Learn More
Aviva Systems Biology Corp ATF7 Antibody - C-terminal region (ARP38869_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38869_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ATF7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ATF7. Alternate names: ATFAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-56826