Learn More
Cayman Chemical ATF7 TrnscrIptIon Factr Mon
SDP
Catalog No. 502949098
Encompass_Preferred
A diterpene alkaloid with diverse biological activities inhibits binding of a-bungarotoxin to rat neuronal a7 nAChRs (IC50 19 UM) repellent against adult red flour beetles in a filter paper assay at 3 mg/ml lowers systolic blood pressure in a rat model of left renal artery ligation-induced hypertension at 75 mg/kg
Catalog No. 50-294-9098 Supplier Cayman Chemical Supplier No. 32756100 UL
A diterpene alkaloid with diverse biological activities inhibits binding of a-bungarotoxin to rat neuronal a7 nAChRs (IC50 19 UM) repellent against adult red flour beetles in a filter paper assay at 3 mg/ml lowers systolic blood pressure in a rat model of left renal artery ligation-induced hypertension at 75 mg/kg