Learn More
MSC ATG Cut-Resistant Glove, ANSI A2, 0.80 mm Palm, 1 ea
Catalog No. 5000794882
Encompass
MaxiFlex Cut gloves are cut-resistant, maintaining a super thin, flexible, dexterous, and comfortable feel. These gloves are DMF-free and free from SVHCs, ensuring user safety and environmental compliance.
  • Comfortable cut protection with innovative fibers and reinforced thumb-forefinger area
  • Features 360° breathability with patented micro-foam coating for optimal hand temperature
  • Outstanding durability, withstanding 18,000 abrasive cycles per millimeter
  • Reduces hand fatigue with a thinner design that mimics the 'hand at rest' position
  • Secure grip through an optimized micro-cup finish
  • Guaranteed cleanliness, dermatologist-tested, washed prior to packaging, and OEKO-TEX® certified
Catalog No. 50-007-94882 Supplier MSC Supplier No. 48089312
MaxiFlex Cut gloves are cut-resistant, maintaining a super thin, flexible, dexterous, and comfortable feel. These gloves are DMF-free and free from SVHCs, ensuring user safety and environmental compliance.
  • Comfortable cut protection with innovative fibers and reinforced thumb-forefinger area
  • Features 360° breathability with patented micro-foam coating for optimal hand temperature
  • Outstanding durability, withstanding 18,000 abrasive cycles per millimeter
  • Reduces hand fatigue with a thinner design that mimics the 'hand at rest' position
  • Secure grip through an optimized micro-cup finish
  • Guaranteed cleanliness, dermatologist-tested, washed prior to packaging, and OEKO-TEX® certified