Learn More
MSC ATG Work Gloves, Micro-Foam Nitrile-Coated, Size Medium, Purple, 1 ea
Catalog No. 5000792922
Encompass
The MaxiDry 56-426 is an ultra-lightweight nitrile glove designed for protection against oils and hazardous chemicals. It offers excellent comfort, flexibility, and a secure grip for precision handling in oily and wet environments.
  • Increased resistance to oils through LIQUItech technology
  • Reduced hand fatigue due to ultra-lightweight synthetic coating and seamless knit liner
  • Secure grip with micro-cup non-slip finish in oily and wet applications
  • Locking cuff ensures a snug fit and prevents slipping
  • Cleanliness guaranteed with HandCare, dermatologically accredited and washed prior to packaging
  • Silicone-free construction for skin-friendliness
  • Seamless string knit shell provides comfort and flexibility
Catalog No. 50-007-92922 Supplier MSC Supplier No. 62882634
The MaxiDry 56-426 is an ultra-lightweight nitrile glove designed for protection against oils and hazardous chemicals. It offers excellent comfort, flexibility, and a secure grip for precision handling in oily and wet environments.
  • Increased resistance to oils through LIQUItech technology
  • Reduced hand fatigue due to ultra-lightweight synthetic coating and seamless knit liner
  • Secure grip with micro-cup non-slip finish in oily and wet applications
  • Locking cuff ensures a snug fit and prevents slipping
  • Cleanliness guaranteed with HandCare, dermatologically accredited and washed prior to packaging
  • Silicone-free construction for skin-friendliness
  • Seamless string knit shell provides comfort and flexibility